Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | taxilin alpha | TXLNA |
IL14, TXLN |
Anti-sense | Chr. 1 - 32179669 - 32198285 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 234 | TMEM234 |
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7 |
2.9kb | Sense | Chr. 1 - 32204807 - 32222371 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map