| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000502901.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009011.hg.1 |
ENST00000523150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006778.hg.1 |
ENST00000499752.2 |
Ensembl |