Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CELF2 antisense RNA 1 | CELF2-AS1 | C10orf31 | -51.1kb | Anti-sense | Chr. 10 - 11316833 - 11344674 on NCBI Build GRCh38 |
| Downstream | USP6 N-terminal like | USP6NL |
RNTRE, TRE2NL, USP6NL-IT1 |
61.7kb | Anti-sense | Chr. 10 - 11460510 - 11611789 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map