Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribonuclease H2 subunit C | RNASEH2C |
AGS3, AYP1 |
-37.8kb | Sense | Chr. 11 - 65717673 - 65720938 on NCBI Build GRCh38 |
| Downstream | adaptor related protein complex 5 beta 1 subunit | AP5B1 |
AP-5, PP1030 |
11.6kb | Sense | Chr. 11 - 65773898 - 65780591 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map