Assay Details
| Gene Symbol: TMCO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011537504.2 | XP_011535806.1 | 9 | 2592 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | defective in cullin neddylation 1 domain containing 2 | DCUN1D2 | C13orf17 | -36.4kb | Anti-sense | Chr. 13 - 113455819 - 113492047 on NCBI Build GRCh38 |
| Downstream | transcription factor Dp-1 | TFDP1 |
DILC, DP1, DRTF1, Dp-1 |
55.5kb | Sense | Chr. 13 - 113584688 - 113641473 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TMCO3
Genomic Map