Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATPase phospholipid transporting 10D (putative) | ATP10D | ATPVD | Sense | Chr. 4 - 47485261 - 47593486 on NCBI Build GRCh38 | |
| Downstream |
corin, serine peptidase |
CORIN |
ATC2, CRN, Lrp4, PEE5, TMPRSS10 |
39.4kb | Anti-sense | Chr. 4 - 47593998 - 47838106 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map