Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
asparaginyl-tRNA synthetase 2, mitochondrial (putative) |
NARS2 |
DFNB94, SLM5, asnRS |
-351.8kb | Sense | Chr. 11 - 78435961 - 78574864 on NCBI Build GRCh38 |
| Downstream | microRNA 708 | MIR708 |
MIRN708, hsa-mir-708 |
473.6kb | Sense | Chr. 11 - 79402022 - 79402109 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map