Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928453 | LOC101928453 | -52.2kb | Anti-sense | Chr. 10 - 14074284 - 14087881 on NCBI Build GRCh38 | |
| Downstream | microRNA 4293 | MIR4293 | 240.7kb | Sense | Chr. 10 - 14383200 - 14383277 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map