Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurotrimin | NTM |
HNT, IGLON2, NTRI |
-434.8kb | Anti-sense | Chr. 11 - 131370326 - 132336822 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC646522 | LOC646522 | 1.0mb | Sense | Chr. 11 - 133783671 - 133810376 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map