Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pecanex homolog 3 (Drosophila) | PCNX3 | PCNXL3 | Anti-sense | Chr. 11 - 65614311 - 65637439 on NCBI Build GRCh38 | |
| Downstream | microRNA 4690 | MIR4690 | 10.7kb | Anti-sense | Chr. 11 - 65636310 - 65636369 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map