Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ret finger protein like 2 | RFPL2 | RNF79 | -111.5kb | Anti-sense | Chr. 22 - 32190435 - 32204844 on NCBI Build GRCh38 |
| Downstream | ret finger protein like 3 | RFPL3 | 32.5kb | Sense | Chr. 22 - 32354885 - 32361161 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map