Assay Details
| Gene Symbol: LOC105371828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126827.1 | - | 1 - 2 | 768 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC440446 | LOC440446 | -97.1kb | Sense | Chr. 17 - 51336715 - 51342571 on NCBI Build GRCh38 | |
| Downstream | carbonic anhydrase 10 | CA10 |
CA-RPX, CARPX, HUCEP-15 |
184.5kb | Anti-sense | Chr. 17 - 51630313 - 52160017 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105371828
Genomic Map