Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ski2 like RNA helicase 2 | SKIV2L2 |
Dob1, KIAA0052, Mtr4, fSAP118 |
-11.8kb | Anti-sense | Chr. 5 - 55307748 - 55425581 on NCBI Build GRCh38 |
| Downstream | microRNA 5687 | MIR5687 | 69.3kb | Sense | Chr. 5 - 55508850 - 55508926 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map