Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DLGAP1 antisense RNA 2 | DLGAP1-AS2 | -22.2kb | Anti-sense | Chr. 18 - 3603737 - 3610089 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105371967 | LOC105371967 | 18.9kb | Anti-sense | Chr. 18 - 3653056 - 3656282 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map