Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis expressed 15 | TEX15 | CT42 | -115.0kb | Sense | Chr. 8 - 30831544 - 30913012 on NCBI Build GRCh38 |
| Downstream | Werner syndrome RecQ like helicase | WRN |
RECQ3, RECQL2, RECQL3 |
3.6kb | Anti-sense | Chr. 8 - 31033262 - 31173761 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map