Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 5690 | MIR5690 | -3.9kb | Sense | Chr. 6 - 35664717 - 35664789 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC285847 | LOC285847 | 55.9kb | Sense | Chr. 6 - 35726762 - 35736947 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map