Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 327 | LINC00327 | NCRNA00327 | -139.8kb | Sense | Chr. 13 - 23469512 - 23487464 on NCBI Build GRCh38 |
| Downstream | mitochondrial intermediate peptidase | MIPEP |
HMIP, MIP |
99.9kb | Anti-sense | Chr. 13 - 23730189 - 23889448 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map