Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC644794 | LOC644794 | -12.2kb | Anti-sense | Chr. 7 - 66904120 - 66906297 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 248 | TMEM248 | C7orf42 | 554.0 | Anti-sense | Chr. 7 - 66921216 - 66958551 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map