Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105369785 | LOC105369785 | -10.5kb | Sense | Chr. 12 - 58087892 - 58093362 on NCBI Build GRCh38 | |
| Downstream | endogenous retrovirus group K member 8 Pol protein-like | LOC105369972 | 217.6kb | Anti-sense | Chr. 12 - 58323535 - 58331743 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map