Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin D2 | CCND2 |
KIAK0002, MPPH3 |
-3.8kb | Sense | Chr. 12 - 4273736 - 4305356 on NCBI Build GRCh38 |
| Downstream | TP53 induced glycolysis regulatory phosphatase | TIGAR |
C12orf5, FR2BP |
10.6kb | Sense | Chr. 12 - 4321193 - 4360028 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map