Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutathione peroxidase 1 | GPX1 |
GPXD, GSHPX1 |
-48.9kb | Sense | Chr. 3 - 49357176 - 49358358 on NCBI Build GRCh38 |
| Downstream | T-cell leukemia translocation altered | TCTA | 3.5kb | Anti-sense | Chr. 3 - 49412206 - 49416476 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map