Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SYNE1 antisense RNA 1 | SYNE1-AS1 | -225.3kb | Anti-sense | Chr. 6 - 152380530 - 152381713 on NCBI Build GRCh38 | |
| Downstream | myc target 1 | MYCT1 | MTLC | 89.5kb | Anti-sense | Chr. 6 - 152697895 - 152747171 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map