Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytohesin 4 | CYTH4 |
CYT4, DJ63G5.1, PSCD4 |
Sense | Chr. 22 - 37282383 - 37315348 on NCBI Build GRCh38 | |
| Downstream | extracellular leucine rich repeat and fibronectin type III domain containing 2 | ELFN2 |
LRRC62, PPP1R29 |
52.1kb | Anti-sense | Chr. 22 - 37340644 - 37427470 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map