Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein L13 | MRPL13 |
L13, L13A, L13mt, RPL13, RPML13 |
-22.9kb | Anti-sense | Chr. 8 - 120395844 - 120445407 on NCBI Build GRCh38 |
| Downstream | syntrophin beta 1 | SNTB1 |
59-DAP, A1B, BSYN2, DAPA1B, SNT2, SNT2B1, TIP-43 |
65.2kb | Anti-sense | Chr. 8 - 120535745 - 120812069 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map