Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosducin like 3 | PDCL3 |
HTPHLP, PHLP2A, PHLP3, VIAF, VIAF1 |
-271.3kb | Sense | Chr. 2 - 100562956 - 100577472 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927142 | LOC101927142 | 122.6kb | Anti-sense | Chr. 2 - 100972648 - 100977161 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map