Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CNTN4 antisense RNA 2 | CNTN4-AS2 | Anti-sense | Chr. 3 - 2110409 - 2144241 on NCBI Build GRCh38 | ||
| Downstream | CNTN4 antisense RNA 1 | CNTN4-AS1 | 910.4kb | Anti-sense | Chr. 3 - 3039661 - 3061145 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map