Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4764 | MIR4764 | -22.8kb | Sense | Chr. 22 - 33436582 - 33436669 on NCBI Build GRCh38 | |
| Downstream | LARGE antisense RNA 1 | LARGE-AS1 | 263.7kb | Anti-sense | Chr. 22 - 33724986 - 33750817 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map