Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gamma-aminobutyric acid type A receptor beta1 subunit | GABRB1 | Anti-sense | Chr. 4 - 47031278 - 47438409 on NCBI Build GRCh38 | ||
| Downstream | COMM domain containing 8 | COMMD8 | 273.3kb | Sense | Chr. 4 - 47450793 - 47463702 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map