Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microtubule associated serine/threonine kinase 2 | MAST2 |
MAST205, MTSSK |
Anti-sense | Chr. 1 - 45786992 - 46036124 on NCBI Build GRCh38 | |
| Downstream | phosphoinositide-3-kinase regulatory subunit 3 | PIK3R3 |
p55, p55-GAMMA, p55PIK |
7.4kb | Sense | Chr. 1 - 46040140 - 46176495 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map