Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoglycerate mutase 1 pseudogene 5 | PGAM1P5 | -45.2kb | Anti-sense | Chr. 12 - 95649255 - 95673999 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105369921 | LOC105369921 | 70.4kb | Sense | Chr. 12 - 95795345 - 95858839 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map