Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GDP-mannose 4,6-dehydratase | GMDS |
GMD, SDR3E1 |
Sense | Chr. 6 - 1623800 - 2245634 on NCBI Build GRCh38 | |
| Downstream | GMDS antisense RNA 1 (head to head) | GMDS-AS1 | 544.8kb | Anti-sense | Chr. 6 - 2245753 - 2413591 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map