Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin protein ligase E3D | UBE3D |
C6orf157, H10BH, UBE2CBP, YJR141W |
Sense | Chr. 6 - 82880834 - 83065841 on NCBI Build GRCh38 | |
| Downstream | dopey family member 1 | DOPEY1 |
DOP1, KIAA1117, dJ202D23.2 |
149.7kb | Anti-sense | Chr. 6 - 83067650 - 83171351 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map