Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S6 kinase A1 | RPS6KA1 |
HU-1, MAPKAPK1A, RSK, RSK1, p90Rsk |
Sense | Chr. 1 - 26529758 - 26575029 on NCBI Build GRCh38 | |
| Downstream | microRNA 1976 | MIR1976 | hsa-mir-1976 | 18.4kb | Sense | Chr. 1 - 26554542 - 26554593 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map