Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 294 | LINC00294 | NCRNA00294 | -11.8kb | Sense | Chr. 11 - 33076150 - 33079454 on NCBI Build GRCh38 |
| Downstream | CSTF3 antisense RNA 1 (head to head) | CSTF3-AS1 | 68.5kb | Sense | Chr. 11 - 33161657 - 33191598 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map