Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | maestro | MRO |
B29, C18orf3 |
-111.6kb | Anti-sense | Chr. 18 - 50795120 - 50825413 on NCBI Build GRCh38 |
| Downstream | elaC ribonuclease Z 1 | ELAC1 | D29 | 28.9kb | Sense | Chr. 18 - 50968017 - 50988120 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map