Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and SCAN domain containing 18 | ZSCAN18 | ZNF447 | -2.6kb | Sense | Chr. 19 - 58083834 - 58118433 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 329 | ZNF329 | 3.6kb | Sense | Chr. 19 - 58126328 - 58155169 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map