Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 710 | LINC00710 | -6.2kb | Anti-sense | Chr. 10 - 10934940 - 10952163 on NCBI Build GRCh38 | |
| Downstream |
CUGBP, Elav-like family member 2 |
CELF2 |
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR |
44.1kb | Sense | Chr. 10 - 10462515 - 11336675 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map