Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8080 | MIR8080 | hsa-mir-8080 | -96.6kb | Anti-sense | Chr. 2 - 79866495 - 79866583 on NCBI Build GRCh38 |
| Downstream | leucine rich repeat transmembrane neuronal 1 | LRRTM1 | 322.5kb | Anti-sense | Chr. 2 - 80301878 - 80304738 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map