Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC653602 | LOC653602 | -211.1kb | Sense | Chr. 2 - 14635091 - 14650819 on NCBI Build GRCh38 | |
| Downstream | neuroblastoma amplified sequence | NBAS |
ILFS2, NAG, SOPH |
302.6kb | Anti-sense | Chr. 2 - 14998067 - 15561348 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map