Assay Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055475.1 | - | 2 - 3 | 535 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytohesin 4 | CYTH4 |
CYT4, DJ63G5.1, PSCD4 |
-24.2kb | Anti-sense | Chr. 22 - 37282383 - 37315348 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100506271 | LOC100506271 | 9.5kb | Sense | Chr. 22 - 37352190 - 37354839 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |

ELFN2
Genomic Map