Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine active site containing 1 | SERAC1 | Sense | Chr. 6 - 158109504 - 158168280 on NCBI Build GRCh38 | ||
| Downstream | general transcription factor IIH subunit 5 | GTF2H5 |
C6orf175, TFB5, TFIIH, TGF2H5, TTD, TTD-A, TTD3, TTDA, bA120J8.2 |
44.6kb | Anti-sense | Chr. 6 - 158168316 - 158199344 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map