Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 1 open reading frame 162 | C1orf162 | Sense | Chr. 1 - 111462694 - 111478512 on NCBI Build GRCh38 | ||
| Downstream | transmembrane and immunoglobulin domain containing 3 | TMIGD3 | AD026 | 10.3kb | Anti-sense | Chr. 1 - 111483348 - 111564003 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map