Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interactor of little elongation complex ELL subunit 2 | ICE2 |
BRCC1, NARG2 |
Sense | Chr. 15 - 60419609 - 60479160 on NCBI Build GRCh38 | |
| Downstream | RORA antisense RNA 1 | RORA-AS1 | 31.6kb | Anti-sense | Chr. 15 - 60479178 - 60630639 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map