Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723809 | LOC102723809 | Anti-sense | Chr. 14 - 64514151 - 64540368 on NCBI Build GRCh38 | ||
| Downstream | heat shock protein family A (Hsp70) member 2 | HSPA2 |
HSP70-2, HSP70-3 |
13.3kb | Sense | Chr. 14 - 64540468 - 64543237 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map