Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 12 open reading frame 54 | C12orf54 |
HSD-29, HSD-30 |
-10.8kb | Sense | Chr. 12 - 48413154 - 48496514 on NCBI Build GRCh38 |
| Downstream | lactalbumin alpha | LALBA | LYZG | 37.8kb | Anti-sense | Chr. 12 - 48567684 - 48571822 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map