Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 905 | LINC00905 | -32.2kb | Anti-sense | Chr. 19 - 16033713 - 16042135 on NCBI Build GRCh38 | |
| Downstream |
RAB8A, member RAS oncogene family |
RAB8A |
MEL, RAB8 |
34.3kb | Anti-sense | Chr. 19 - 16111680 - 16133635 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map