Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507557 | LOC100507557 | -8.5kb | Anti-sense | Chr. 6 - 145734869 - 145886585 on NCBI Build GRCh38 | |
| Downstream | glutamate metabotropic receptor 1 | GRM1 |
GPRC1A, MGLU1, MGLUR1, PPP1R85, SCAR13 |
67.3kb | Anti-sense | Chr. 6 - 146027634 - 146437598 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map