Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heart development protein with EGF like domains 1 | HEG1 |
HEG, MST112, MSTP112 |
Sense | Chr. 3 - 124965710 - 125055958 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 12 member 8 | SLC12A8 | CCC9 | 99.3kb | Sense | Chr. 3 - 125082636 - 125212765 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map