Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger MYND-type containing 8 | ZMYND8 |
PRKCBP1, PRO2893, RACK7 |
-177.5kb | Anti-sense | Chr. 20 - 47209214 - 47357780 on NCBI Build GRCh38 |
| Downstream | sulfatase 2 | SULF2 | HSULF-2 | 121.0kb | Anti-sense | Chr. 20 - 47654584 - 47786629 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map