Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FKBP1A-SDCBP2 readthrough (NMD candidate) | FKBP1A-SDCBP2 | Anti-sense | Chr. 20 - 1309909 - 1393172 on NCBI Build GRCh38 | ||
| Downstream | SDCBP2 antisense RNA 1 | SDCBP2-AS1 | 976.0 | Sense | Chr. 20 - 1325343 - 1378735 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map